Ethyl 4-bromo-3-hydroxybenzoate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR949729-1G OR949729-250MG OR949729-5G
APOSOR949729-1GEA 218 DKK
APOSOR949729-1G APOSOR949729-250MG APOSOR949729-5G
Ethyl 4-bromo-3-hydroxybenzoate 98%
Ethyl 4-bromo-3-hydroxybenzoate
Formel: C₉H₉BrO₃
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09038129
CAS-nummer: 33141-66-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR