5-Phenyl-1H-1,2,3-triazole 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR949726-1G OR949726-5G
APOSOR949726-1GEA 1640 DKK
APOSOR949726-1G APOSOR949726-5G
5-Phenyl-1H-1,2,3-triazole 95%
5-Phenyl-1H-1,2,3-triazole
Formel: C₈H₇N₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06739070
CAS-nummer: 1680-44-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR