Ethyl 3-oxopiperidine-1-carboxylate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR947016-1G OR947016-5G
APOSOR947016-1GEA 1040 DKK
APOSOR947016-1G APOSOR947016-5G
Ethyl 3-oxopiperidine-1-carboxylate 95%
Ethyl 3-oxopiperidine-1-carboxylate
Formel: C₈H₁₃NO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD15144251
CAS-nummer: 61995-19-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR