2-(Cyclopentyloxy)benzaldehyde 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR946444-1G
OR946444-250MG
OR946444-5G
APOSOR946444-1GEA
2030
DKK
APOSOR946444-1G
APOSOR946444-250MG
APOSOR946444-5G
2-(Cyclopentyloxy)benzaldehyde 97%
2-(Cyclopentyloxy)benzaldehyde
Formel:
C₁₂H₁₄O₂ Molekylvægt: 190.238 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04132910 CAS-nummer: 145742-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...