4-(Chloromethyl)-N-ethylbenzamide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR946358-10G OR946358-1G OR946358-5G
APOSOR946358-10GEA 4860 DKK
APOSOR946358-10G APOSOR946358-1G APOSOR946358-5G
4-(Chloromethyl)-N-ethylbenzamide 98%
4-(Chloromethyl)-N-ethylbenzamide
Formel: C₁₀H₁₂ClNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11208368
CAS-nummer: 852157-71-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR