5-Methyl-D-tryptophan 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR946064-1G
OR946064-250MG
OR946064-500MG
APOSOR946064-1GEA
6540
DKK
APOSOR946064-1G
APOSOR946064-250MG
APOSOR946064-500MG
5-Methyl-D-tryptophan 97%
5-Methyl-D-tryptophan
Formel:
C₁₂H₁₄N₂O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09841957 CAS-nummer: 99295-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...