N,N,2-Trimethylquinolin-6-amine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR945425-1G OR945425-250MG
APOSOR945425-1GEA 0 DKK
APOSOR945425-1G APOSOR945425-250MG
N,N,2-Trimethylquinolin-6-amine 95%
N,N,2-Trimethylquinolin-6-amine
Formel: C₁₂H₁₄N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00089173
CAS-nummer: 92-99-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR