3-(tert-Butyl)-2-hydroxybenzonitrile 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR942363-1G OR942363-250MG
APOSOR942363-1GEA 523 DKK
APOSOR942363-1G APOSOR942363-250MG
3-(tert-Butyl)-2-hydroxybenzonitrile 95%
3-(tert-Butyl)-2-hydroxybenzonitrile
Formel: C₁₁H₁₃NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16999197
CAS-nummer: 340131-70-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR