2-Hydroxyphenylcyclopropyl ketone 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Cyclopropyl(2-hydroxyphenyl)methanone
OR941165-1G
OR941165-250MG
APOSOR941165-1GEA
622
DKK
APOSOR941165-1G
APOSOR941165-250MG
2-Hydroxyphenylcyclopropyl ketone 95%
2-Hydroxyphenylcyclopropyl ketone
Formel:
C₁₀H₁₀O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD24703561 CAS-nummer: 128405-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...