5-Hydroxy-3-methylindole 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR940620-1G
OR940620-250MG
APOSOR940620-1GEA
1480
DKK
APOSOR940620-1G
APOSOR940620-250MG
5-Hydroxy-3-methylindole 95%
5-Hydroxy-3-methylindole
Formel:
C₉H₉NO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03095210 CAS-nummer: 1125-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...