2-Chloro-5-phenylpyrimidine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR938765-1G OR938765-250MG
APOSOR938765-1GEA 1330 DKK
APOSOR938765-1G APOSOR938765-250MG
2-Chloro-5-phenylpyrimidine 95%
2-Chloro-5-phenylpyrimidine
Formel: C₁₀H₇ClN₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04035567
CAS-nummer: 22536-62-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR