(3-O-Tolyl-isoxazol-5-yl)-methanol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-(Hydroxymethyl)-3-(o-tolyl)isoxazole

OR932964-1G OR932964-250MG
APOSOR932964-1GEA 1820 DKK
APOSOR932964-1G APOSOR932964-250MG
(3-O-Tolyl-isoxazol-5-yl)-methanol 95%
(3-O-Tolyl-isoxazol-5-yl)-methanol
Formel: C₁₁H₁₁NO₂
Molekylvægt: 189.211 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06738508
CAS-nummer: 885273-56-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR