Methyl 4-bromo-3-cyanobenzoate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR932422-1G OR932422-5G
APOSOR932422-1GEA 1140 DKK
APOSOR932422-1G APOSOR932422-5G
Methyl 4-bromo-3-cyanobenzoate 97%
Methyl 4-bromo-3-cyanobenzoate
Formel: C₉H₆BrNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18398742
CAS-nummer: 1232424-44-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR