2-Methylaminoethyl chloride hydrochloride 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR932350-10G
OR932350-1G
OR932350-25G
OR932350-100G
APOSOR932350-10GEA
513
DKK
APOSOR932350-10G
APOSOR932350-1G
APOSOR932350-25G
APOSOR932350-100G
2-Methylaminoethyl chloride hydrochloride 97%
2-Methylaminoethyl chloride hydrochloride
Formel:
C₃H₈ClN·ClH Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00050517 CAS-nummer: 4535-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...