4-Methylpent-4-enoic acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR930895-1G
OR930895-250MG
APOSOR930895-1GEA
2960
DKK
APOSOR930895-1G
APOSOR930895-250MG
4-Methylpent-4-enoic acid 95%
4-Methylpent-4-enoic acid
Formel:
C₆H₁₀O₂ Molekylvægt: 114.142 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06208588 CAS-nummer: 1001-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...