4-(Diethylamino)-2-methylbenzaldehyde 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR930554-1G OR930554-250MG
APOSOR930554-1GEA 1600 DKK
APOSOR930554-1G APOSOR930554-250MG
4-(Diethylamino)-2-methylbenzaldehyde 95%
4-(Diethylamino)-2-methylbenzaldehyde
Formel: C₁₂H₁₇NO
Molekylvægt: 191.269 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06800551
CAS-nummer: 92-14-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR