2,2-Dimethyl-3-oxo-butyric acid ethyl ester 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR929960-1G OR929960-5G
APOSOR929960-1GEA 1810 DKK
APOSOR929960-1G APOSOR929960-5G
2,2-Dimethyl-3-oxo-butyric acid ethyl ester 95%
2,2-Dimethyl-3-oxo-butyric acid ethyl ester
Formel: C₈H₁₄O₃
Molekylvægt: 158.195 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06658443
CAS-nummer: 597-04-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR