4-(4-Acetyl-1-piperazinyl)phenylboronic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR929583-1G OR929583-250MG
APOSOR929583-1GEA 3140 DKK
APOSOR929583-1G APOSOR929583-250MG
4-(4-Acetyl-1-piperazinyl)phenylboronic acid 95%
4-(4-Acetyl-1-piperazinyl)phenylboronic acid
Formel: C₁₂H₁₇BN₂O₃
Opbevaringstemperatur: Fryser
MDL Number: MFCD21332885
CAS-nummer: 1384954-82-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR