3-Methoxypyrazine-2-carboxylic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR929007-1G OR929007-250MG
APOSOR929007-1GEA 644 DKK
APOSOR929007-1G APOSOR929007-250MG
3-Methoxypyrazine-2-carboxylic acid 98%
3-Methoxypyrazine-2-carboxylic acid
Formel: C₆H₆N₂O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10697766
CAS-nummer: 40155-47-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR