5-Aminobenzo[b]furan 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR9276-1G OR9276-5G OR9276-250MG
APOSOR9276-1GEA 1340 DKK
APOSOR9276-1G APOSOR9276-5G APOSOR9276-250MG
5-Aminobenzo[b]furan 97%
5-Aminobenzo[b]furan
Formel: C₈H₇NO
Opbevaringstemperatur: Fryser
MDL Number: MFCD04125391
CAS-nummer: 58546-89-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR