3-Methoxy-5-methyl-isoxazole 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR927106-1G OR927106-250MG
APOSOR927106-1GEA 4860 DKK
APOSOR927106-1G APOSOR927106-250MG
3-Methoxy-5-methyl-isoxazole 95%
3-Methoxy-5-methyl-isoxazole
Formel: C₅H₇NO₂
Molekylvægt: 113.115 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22565666
CAS-nummer: 16864-45-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR