1-Isopropyl-2,3-dihydro-1H-indole-5-carbaldehyde 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Isopropylindoline-5-carbaldehyde

OR926947-1G OR926947-250MG
APOSOR926947-1GEA 2550 DKK
APOSOR926947-1G APOSOR926947-250MG
1-Isopropyl-2,3-dihydro-1H-indole-5-carbaldehyde 95%
1-Isopropyl-2,3-dihydro-1H-indole-5-carbaldehyde
Formel: C₁₂H₁₅NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07186368
CAS-nummer: 876710-70-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR