N-Boc-endo-3-aminotropane 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR926114-1G
OR926114-5G
APOSOR926114-1GEA
0
DKK
APOSOR926114-1G
APOSOR926114-5G
N-Boc-endo-3-aminotropane 97%
N-Boc-endo-3-aminotropane
Formel:
C₁₂H₂₂N₂O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD18428077 CAS-nummer: 207405-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...