4-Chloro-5-propylpyrimidine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR925515-1G OR925515-250MG
APOSOR925515-1GEA 4520 DKK
APOSOR925515-1G APOSOR925515-250MG
4-Chloro-5-propylpyrimidine 98%
4-Chloro-5-propylpyrimidine
Formel: C₇H₉ClN₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09971219
CAS-nummer: 25199-00-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR