Ethyl 5-chloropyrimidine-2-carboxylate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR924082-1G OR924082-250MG
APOSOR924082-1GEA 6050 DKK
APOSOR924082-1G APOSOR924082-250MG
Ethyl 5-chloropyrimidine-2-carboxylate 95%
Ethyl 5-chloropyrimidine-2-carboxylate
Formel: C₇H₇ClN₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16610953
CAS-nummer: 1227571-82-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR