(S)-3-Amino-1-methyl-piperidine dihydrochloride 96%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR924007-1G
OR924007-250MG
APOSOR924007-1GEA
506
DKK
APOSOR924007-1G
APOSOR924007-250MG
(S)-3-Amino-1-methyl-piperidine dihydrochloride 96%
(S)-3-Amino-1-methyl-piperidine dihydrochloride
Formel:
C₆H₁₄N₂·2ClH Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD20926142 CAS-nummer: 1157849-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...