Cyclohexane-1,4-dicarbonitrile 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR923541-1G OR923541-5G
APOSOR923541-1GEA 1730 DKK
APOSOR923541-1G APOSOR923541-5G
Cyclohexane-1,4-dicarbonitrile 95%
Cyclohexane-1,4-dicarbonitrile
Formel: C₈H₁₀N₂
Molekylvægt: 134.178 g/mol
Smeltepunkt: 57…61 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00021283
CAS-nummer: 10534-13-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR