Isobutylsulphamoyl chloride 95%
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR922364-1G OR922364-250MG
APOSOR922364-1GEA 3100 DKK
APOSOR922364-1G APOSOR922364-250MG
Isobutylsulphamoyl chloride 95%
Isobutylsulphamoyl chloride
Formel: C₄H₁₀ClNO₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11520374
CAS-nummer: 26118-68-3