(S)-Fmoc-2-amino-3-ethyl-pentanoic acid 99%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR921460-100MG OR921460-1G OR921460-250MG
APOSOR921460-100MGEA 3220 DKK
APOSOR921460-100MG APOSOR921460-1G APOSOR921460-250MG
(S)-Fmoc-2-amino-3-ethyl-pentanoic acid 99%
(S)-Fmoc-2-amino-3-ethyl-pentanoic acid
Formel: C₂₂H₂₅NO₄
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11226813
CAS-nummer: 1310680-47-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR