7-Bromobicyclo[4.2.0]octa-1,3,5-triene 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR918719-1G OR918719-250MG
APOSOR918719-1GEA 1260 DKK
APOSOR918719-1G APOSOR918719-250MG
7-Bromobicyclo[4.2.0]octa-1,3,5-triene 98%
7-Bromobicyclo[4.2.0]octa-1,3,5-triene
Formel: C₈H₇Br
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD17976891
CAS-nummer: 21120-91-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR