Methyl 2,2,2-trimethoxyacetate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR918584-1G OR918584-25G OR918584-5G
APOSOR918584-1GEA 452 DKK
APOSOR918584-1G APOSOR918584-25G APOSOR918584-5G
Methyl 2,2,2-trimethoxyacetate 95%
Methyl 2,2,2-trimethoxyacetate
Formel: C₆H₁₂O₅
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09701200
CAS-nummer: 18370-95-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR