4-Chloro-2-oxoindoline-3-carbaldehyde 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR917736-1G OR917736-250MG
APOSOR917736-1GEA 6050 DKK
APOSOR917736-1G APOSOR917736-250MG
4-Chloro-2-oxoindoline-3-carbaldehyde 95%
4-Chloro-2-oxoindoline-3-carbaldehyde
Formel: C₉H₆ClNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20502958
CAS-nummer: 23872-23-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR