6-Bromo-D-tryptophan 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR916897-1G OR916897-250MG OR916897-500MG
APOSOR916897-1GEA 7360 DKK
APOSOR916897-1G APOSOR916897-250MG APOSOR916897-500MG
6-Bromo-D-tryptophan 97%
6-Bromo-D-tryptophan
Formel: C₁₁H₁₁BrN₂O₂
Molekylvægt: 283.121 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD17214434
CAS-nummer: 496930-10-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR