2-Isopropyl-2H-3-thiazole 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR914221-1G OR914221-5G
APOSOR914221-1GEA 1260 DKK
APOSOR914221-1G APOSOR914221-5G
2-Isopropyl-2H-3-thiazole 98%
2-Isopropyl-2H-3-thiazole
Formel: C₆H₉NS
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02181138
CAS-nummer: 15679-10-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR