Butane-1-sulphonamide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR911092-10G OR911092-1G OR911092-5G
APOSOR911092-10GEA 0 DKK
APOSOR911092-10G APOSOR911092-1G APOSOR911092-5G
Butane-1-sulphonamide 98%
Butane-1-sulphonamide
Formel: C₄H₁₁NO₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00465180
CAS-nummer: 3144-04-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR