3-(2-Cyanophenyl)-N,N-dimethylbenzamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR910837-1G OR910837-5G
APOSOR910837-1GEA 1700 DKK
APOSOR910837-1G APOSOR910837-5G
3-(2-Cyanophenyl)-N,N-dimethylbenzamide
3-(2-Cyanophenyl)-N,N-dimethylbenzamide
Formel: C₁₆H₁₄N₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD21609619
CAS-nummer: 1365271-34-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR