3-formyl-1H-indole-6-carboxylic acid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR910510-1G
OR910510-250MG
APOSOR910510-1GEA
3870
DKK
APOSOR910510-1G
APOSOR910510-250MG
3-formyl-1H-indole-6-carboxylic acid 97%
3-formyl-1H-indole-6-carboxylic acid
Formel:
C₁₀H₇NO₃ Molekylvægt: 189.167 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06656877 CAS-nummer: 887576-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...