tert-butyl N-ethenylcarbamate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR909976-1G OR909976-250MG
APOSOR909976-1GEA 183 DKK
APOSOR909976-1G APOSOR909976-250MG
tert-butyl N-ethenylcarbamate 95%
tert-butyl N-ethenylcarbamate
Formel: C₇H₁₃NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081075
CAS-nummer: 7150-72-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR