2-Methoxy-4-pyrrolidinopyrimidine 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR909525-1G
OR909525-5G
APOSOR909525-1GEA
1640
DKK
APOSOR909525-1G
APOSOR909525-5G
2-Methoxy-4-pyrrolidinopyrimidine 97%
2-Methoxy-4-pyrrolidinopyrimidine
Formel:
C₉H₁₃N₃O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD22383693 CAS-nummer: 1393442-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...