2-(Dimethoxymethyl)pyridine-5-boronic acid, pinacol ester 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-(Dimethoxymethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
OR908949-250MG
OR908949-1G
OR908949-5G
APOSOR908949-250MGEA
556
DKK
APOSOR908949-250MG
APOSOR908949-1G
APOSOR908949-5G
2-(Dimethoxymethyl)pyridine-5-boronic acid, pinacol ester 98%
2-(Dimethoxymethyl)pyridine-5-boronic acid, pinacol ester
Formel:
C₁₄H₂₂BNO₄ Molekylvægt: 279.14 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD16038216 CAS-nummer: 1150632-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...