2-Cyclopropyloxybenzeneboronic acid, pinacol ester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(2-Cyclopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR908570-1G OR908570-250MG
APOSOR908570-1GEA 2270 DKK
APOSOR908570-1G APOSOR908570-250MG
2-Cyclopropyloxybenzeneboronic acid, pinacol ester
2-Cyclopropyloxybenzeneboronic acid, pinacol ester
Formel: C₁₅H₂₁BO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16994462
CAS-nummer: 1119090-11-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR