3-tert-Butyl-4-hydroxybenzoic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-(tert-Butyl)-4-hydroxybenzoic acid

OR908194-1G OR908194-5G
APOSOR908194-1GEA 665 DKK
APOSOR908194-1G APOSOR908194-5G
3-tert-Butyl-4-hydroxybenzoic acid 97%
3-tert-Butyl-4-hydroxybenzoic acid
Formel: C₁₁H₁₄O₃
Molekylvægt: 194.227 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016535
CAS-nummer: 66737-88-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR