Methyl 4-thiocyanatobutanoate 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR907804-1G
OR907804-5G
APOSOR907804-1GEA
1260
DKK
APOSOR907804-1G
APOSOR907804-5G
Methyl 4-thiocyanatobutanoate 97%
Methyl 4-thiocyanatobutanoate
Formel:
C₆H₉NO₂S Molekylvægt: 159.206 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD15143541 CAS-nummer: 79203-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...