Methyl d-4-chlorophenylglycinate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR905392-1G OR905392-250MG
APOSOR905392-1GEA 3270 DKK
APOSOR905392-1G APOSOR905392-250MG
Methyl d-4-chlorophenylglycinate 95%
Methyl d-4-chlorophenylglycinate
Formel: C₉H₁₀ClNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07784110
CAS-nummer: 43189-43-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR