(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR905340-1G OR905340-250MG
APOSOR905340-1GEA 5130 DKK
APOSOR905340-1G APOSOR905340-250MG
(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid 95%
(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid
Formel: C₇H₉ClN₂O₂
Molekylvægt: 188.612 g/mol
MDL Number: MFCD01114908
CAS-nummer: 374913-86-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR