3-(Isocyanomethyl)pyridine 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR904838-1G
OR904838-250MG
APOSOR904838-1GEA
5130
DKK
APOSOR904838-1G
APOSOR904838-250MG
3-(Isocyanomethyl)pyridine 95%
3-(Isocyanomethyl)pyridine
Formel:
C₇H₆N₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02664552 CAS-nummer: 58644-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...