Ethyl benzofurazan-5-carboxylate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR8306-1G
OR8306-5G
OR8306-10G
APOSOR8306-1GEA
765
DKK
APOSOR8306-1G
APOSOR8306-5G
APOSOR8306-10G
Ethyl benzofurazan-5-carboxylate
Ethyl benzofurazan-5-carboxylate
Formel:
C₉H₈N₂O₃ Smeltepunkt: 44…46 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00276586 CAS-nummer: 36389-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...