4,5,6-Triaminopyrimidine sulphate 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Pyrimidine-4,5,6-triamine sulphate
OR8231-1G
OR8231-5G
APOSOR8231-1GEA
850
DKK
APOSOR8231-1G
APOSOR8231-5G
4,5,6-Triaminopyrimidine sulphate 95%
4,5,6-Triaminopyrimidine sulphate
Formel:
C₄H₇N₅·H₂SO₄ Smeltepunkt: >300 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00012789 CAS-nummer: 49721-45-1 EINECS: 256-446-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...