1-(Biphenyl-4-yl)piperazine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Biphenyl-4-yl-piperazine, 4-(Piperazin-1-yl)biphenyl

OR8173-1G OR8173-5G
APOSOR8173-1GEA 1530 DKK
APOSOR8173-1G APOSOR8173-5G
1-(Biphenyl-4-yl)piperazine
1-(Biphenyl-4-yl)piperazine
Formel: C₁₆H₁₈N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02093529
CAS-nummer: 180698-19-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR