Ethyl 3-(4-hydroxyphenyl)propanoate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethyl 3-(4-hydroxyphenyl)propionate, 4-(3-Ethoxy-3-oxoprop-1-yl)phenol

OR8094-1G OR8094-5G
APOSOR8094-1GEA 186 DKK
APOSOR8094-1G APOSOR8094-5G
Ethyl 3-(4-hydroxyphenyl)propanoate 95%
Ethyl 3-(4-hydroxyphenyl)propanoate
Formel: C₁₁H₁₄O₃
Molekylvægt: 194.227 g/mol
Smeltepunkt: 40…42 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00020209
CAS-nummer: 23795-02-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR